Learn More
Carbosynth LLC. MOCAc-Ala-Pro-Ala-Lys-Phe-Phe-Arg-Leu-Lys(Dnp)-NH2 (1mg vial), 1ea, MW: 1458.6, Molecular formula: C71H95N17O17, CAS# 177337-81-4
SDP
Supplier:  Carbosynth LLC. SMO3216V1MG
Encompass_Preferred
Synonyms: (7-Methoxycoumarin-4-yl)Acetyl-L-Alanyl-L-Prolyl-L-Alanyl-LLysyl-L-Phenylalanyl-L-Phenylalanyl-L-Arginyl-L-Leucyl- [Nγ-(2,4-Dinitrophenyl)-L-Lysine] Amide; Mca-Ala-Pro-Ala-Lys-Phe-Phe-Arg-Leu-Lys(Dnp)-NH2 , Mca-APAKFFRLK(Dnp)-NH2, Purity: , Grade A, Application: Fluorescence-Quenching Substrate for Proteinase A / Pepsin, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: H. Kondo, Y. Shibano, T. Amachi, N. Cronin, K. Oda, and B.M. Dunn, J. Biochem., 124, 141 (1998).
Catalog No. 50-168-7132