Learn More
Bachem Mca-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH2
SDP
Catalog No. 50260473
Encompass_Preferred
Mca-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH2 Trifluoroacetate, 158584-09-9, C78H110N22O20, Mr 1675.87, 1mg. NFF-3. The FRET substrate Mca-RPKPVE-Nva-WRK(Dnp) amide was hydrolyzed rapidly by stromelysin 1 (MMP-3) with kcat/Km= 218000 M-1s-1 and very slowly by gelatinase B (MMP-9) (kcat/Km= 10100 M-1s-1). There was no hydrolysis by MMP-1 and MMP-2.
Catalog No. 50-260-473 Supplier Bachem Supplier No. 4030842.0001
Add to Cart
Add to Cart
Mca-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH2 Trifluoroacetate, 158584-09-9, C78H110N22O20, Mr 1675.87, 1mg. NFF-3. The FRET substrate Mca-RPKPVE-Nva-WRK(Dnp) amide was hydrolyzed rapidly by stromelysin 1 (MMP-3) with kcat/Km= 218000 M-1s-1 and very slowly by gelatinase B (MMP-9) (kcat/Km= 10100 M-1s-1). There was no hydrolysis by MMP-1 and MMP-2.