Learn More
CPC Scientific MCA-Gly-Ala-Lys(Ac)-Arg-His-Arg-Lys-Val-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST008A
Encompass
This peptide is the substrate for class I histone deacetylase HDAC 3.SEQUENCE: MCA-Gly-Ala-Lys(Ac)-Arg-His-Arg-Lys-Val-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-GAK(Ac)-RHRKV-NH2MOLECULAR FORMULA: C54H85N19O13MOLECULAR WEIGHT: 1208.39STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [1926163-18-9]SYNONYMS: RESEARCH AREA: MiscellaneousREFERENCES:A.Lahm et al., Proc. Natl. Acad. Sci. USA, 104, 17335 (2007)SKU(s): HIST-008A, HIST-008B
Catalog No. 50-210-9436