Learn More
CPC Scientific MCA-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-DArg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110495
Encompass
Cathespin D and E substrate.SEQUENCE: MCA-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-DArg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-GKPILFFRL-Lys(Dnp)-DArg-NH2MOLECULAR FORMULA: C85H122N22O19MOLECULAR WEIGHT: 1756.04STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [839730-93-7]SYNONYMS: Mca-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(Dnp)-D-Arg-NHRESEARCH AREA: ApoptosisCITATIONS: Wendt, George, et al. BioRxiv (2020).REFERENCES:Y. Yasuda et al., J. Biochem., 125, 1137 (1999)SKU(s): SUBS-017A, SUBS-017B
Catalog No. 50-211-0495 Supplier CPC Scientific Supplier No. SUBS017B
Cathespin D and E substrate.SEQUENCE: MCA-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(DNP)-DArg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Mca-GKPILFFRL-Lys(Dnp)-DArg-NH2MOLECULAR FORMULA: C85H122N22O19MOLECULAR WEIGHT: 1756.04STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [839730-93-7]SYNONYMS: Mca-Gly-Lys-Pro-Ile-Leu-Phe-Phe-Arg-Leu-Lys(Dnp)-D-Arg-NHRESEARCH AREA: ApoptosisCITATIONS: Wendt, George, et al. BioRxiv (2020).REFERENCES:Y. Yasuda et al., J. Biochem., 125, 1137 (1999)SKU(s): SUBS-017A, SUBS-017B