Learn More
CPC Scientific MCA-Leu-Glu-Val-Asp-Gly-Trp-Lys(DNP)-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CASP068B
Encompass
Fluorogenic resonance energy transfer substrate for caspase-4 (ICH-2) that exhibits an excitation wavelength of 325 nm and an emission wavelength of 392 nm.ONE-LETTER SEQUENCE: Mca-LEVDGWK(DNP)-NH2MOLECULAR FORMULA: C57H70N12O19MOLECULAR WEIGHT:1227.24STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [400727-81-3], RESEARCH AREA: ApoptosisREFERENCES: R.V.Talanian et al., J. Biol. Chem., 272, 9677 (1997)
Catalog No. 50-210-9044