Learn More
Medchemexpress LLC Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp) | 185698-23-1 | MFCD01318850 | 99.3% | 892.91 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP37925MG
Encompass_Preferred
Mca-Pro-Leu-Gly-Pro-D-Lys(Dnp) is a FRET substrate of Thimet oligopeptidase, used for determining its activity. This product is also widely utilized in research areas such as peptide synthesis, bioconjugation, drug delivery systems, neuroscience, and diagnostic applications.
  • FRET substrate for Thimet oligopeptidase
  • Used for the determination of Thimet oligopeptidase activity
  • Serves as a building block in peptide synthesis
  • Utilized in bioconjugation processes
  • Can be incorporated into drug delivery systems
  • Allows for the study of neuropeptides
  • Explored in diagnostic applications
Catalog No. 50-003-96725