Learn More
Carbosynth LLC. MOCAc-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH2 (1mg vial) [NFF-3], 1ea, MW: 1675.8, Molecular formula: C78H110N22O20, CAS# 158584-09-9
SDP
Supplier:  Carbosynth LLC. SMO3168V1MG
Encompass_Preferred
Synonyms: NFF-3, (7-Methoxycoumarin-4-yl)acetyl-L-Arginyl-L-Prolyl-L-Lysyl-L-Prolyl-L-Valyl-L-Glutamyl-L-Norvalyl-L-Tryptophyl-L-Arginyl-[Nγ-(2,4-nitrophenyl)-L-Lysine] Amide, Mca-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys(Dnp)-NH2, Mca-RPKPVE-Nva-WRK(Dnp)-NH2, Purity: , Grade B, Application: Fluorescence-Quenching Substrate for Matrix Metalloproteinase-3 (Stromelysin 1), Storage Temperature: 2-10°C, References: H. Nagase, C.G. Fields, and G.B. Fields, J. Biol. Chem., 269, 20952 (1994).
Catalog No. 50-168-7129