Learn More
Medchemexpress LLC Mca-SEVNLDAEFK(Dnp)-NH2 | 1802078-32-5 | 98.8% | 1532.52 | 1 MG
SDP
Catalog No. 5000398133
Encompass_Preferred
Mca-SEVNLDAEFK(Dnp)-NH2 contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2,4-dinitrophenyl group. It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence, such as can be used to measure the activity of BACE-1.
  • Peptide and derivatives
  • Inhibitors and substrates
  • Enzyme substrates
  • Fluorescent dye
  • Fluorescent substrate for peptidases
  • Measures activity of BACE-1
  • Inhibitor
Catalog No. 50-003-98133 Supplier Medchemexpress LLC Supplier No. HYP49201MG
Mca-SEVNLDAEFK(Dnp)-NH2 contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2,4-dinitrophenyl group. It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence, such as can be used to measure the activity of BACE-1.
  • Peptide and derivatives
  • Inhibitors and substrates
  • Enzyme substrates
  • Fluorescent dye
  • Fluorescent substrate for peptidases
  • Measures activity of BACE-1
  • Inhibitor