Learn More
Tecan 100L MCA96 Tip NFil
Catalog No. NC0471753
Encompass_Preferred
Genuine Tecan 100 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
This item is not returnable. View return policy
Catalog No. NC0471753 Supplier Tecan Supplier No. 30038611
Genuine Tecan 100 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.