Learn More
Enzo Life Sciences Mca-YVADAPK(Dnp)-OH (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260023M001
Encompass_Preferred
Specific, highly fluorescent substrate for caspase-1. Also substrate for caspase-4. Alternative name: Caspase-1 substrate (fluorogenic). Formula: C53H64N10O19. MW: 1145.1. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO or 80% acetic acid (1mg/ml, with agitation). Long Term Storage: -20°C.
Catalog No. 50-200-8229