Learn More
Tecan 50L MCA96 Tip NF Nest
Catalog No. NC9085687
Encompass_Preferred
Genuine Tecan 50 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, nested 8 stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 5 tip boxes x 96 tips/rack, 3,840 tips/sales unit. Nested disposable tips offer improved throughput and walk way time due to nested format allowing more tips per carrier. They also give you increased flexibility on the work deck due to ANSI format and are more environmentally friendly, through significant weigh and waste reduction. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.
This item is not returnable. View return policy
Catalog No. NC9085687 Supplier Tecan Supplier No. 30038609
Genuine Tecan 50 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, nested 8 stack, non-filtered. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 5 tip boxes x 96 tips/rack, 3,840 tips/sales unit. Nested disposable tips offer improved throughput and walk way time due to nested format allowing more tips per carrier. They also give you increased flexibility on the work deck due to ANSI format and are more environmentally friendly, through significant weigh and waste reduction. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.