Learn More
Cayman Chemical McaAVLQSGFRK DnpKNH2tr 10mg
SDP
Catalog No. 502915455
Encompass_Preferred
A fluorogenic substrate for SARS-CoV Mpro; Mca is released upon cleavage by SARS-CoV Mpro and its fluorescence can be used to quantify SARS-CoV Mpro activity; ex/em = 328/420 nm, respectively
Catalog No. 50-291-5455 Supplier Cayman Chemical Supplier No. 3700210 MG
A fluorogenic substrate for SARS-CoV Mpro; Mca is released upon cleavage by SARS-CoV Mpro and its fluorescence can be used to quantify SARS-CoV Mpro activity; ex/em = 328/420 nm, respectively