Learn More
Cayman Chemical McaKPLGLDap DnpARNH2tr 10mg
SDP
Catalog No. 502914826
Encompass_Preferred
A fluorogenic substrate for MMPs; Mca is released upon cleavage by MMPs and its fluorescence can be used to quantify MMP activity; ex/em = 328/420 nm, respectively
Catalog No. 50-291-4826 Supplier Cayman Chemical Supplier No. 3696510 MG
A fluorogenic substrate for MMPs; Mca is released upon cleavage by MMPs and its fluorescence can be used to quantify MMP activity; ex/em = 328/420 nm, respectively