Learn More
Aviva Systems Biology Corp MCAM Antibody - middle region (ARP63483_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63483_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCAM. It was validated on Western Blot. . Alternate names: CD146, MUC18ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-287-983