Learn More
ACROBiosystems ACROBiosystems Human MCAM / CD146 Protein, His Tag
Supplier:  ACROBiosystems CD6H5229100UG
Encompass_Preferred
Recombinant Protein;100UG;Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession # NP_006491.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6440