Learn More
ACROBiosystems Recombinant Protein;Human MCAM / CD146 Protein;HEK293;1MG;CD6,MCAM,CD146,MUC18
Catalog No. 501657507
Encompass_Preferred
Recombinant Protein;Human MCAM / CD146 Protein;1MG;Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession # NP_006491.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.3 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7507 Supplier ACROBiosystems Supplier No. CD6H52291MG
Recombinant Protein;Human MCAM / CD146 Protein;1MG;Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession # NP_006491.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.3 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.