Learn More
Doccol Corporation 6-0 medium A MCAO suture L12, 10/pk
SDP
Catalog No. NC1773962
Encompass
Silicone rubber-coated monofilament for MCAO model. Filament size 6-0, diameter 0.09-0.11 mm, length 20 mm; diameter with coating 0.22 +/- 0.02 mm, suitable for MCAO models in animals within body weight range 25 +/- 2 grams; coating length 1-2 mm, more suitable for skilled surgeons doing longer (>60 min) MCAO followed by reperfusion. Pack of 10 filaments.
Catalog No. NC1773962 Supplier Doccol Corporation Supplier No. 602212PK10
Silicone rubber-coated monofilament for MCAO model. Filament size 6-0, diameter 0.09-0.11 mm, length 20 mm; diameter with coating 0.22 +/- 0.02 mm, suitable for MCAO models in animals within body weight range 25 +/- 2 grams; coating length 1-2 mm, more suitable for skilled surgeons doing longer (>60 min) MCAO followed by reperfusion. Pack of 10 filaments.