Learn More
ATCC McA-RH7777; Hepatoma; Rat (Rattus norvegicus)
Supplier:  ATCC CRL1601
Encompass
McA-RH7777 is a cell exhibiting epithelial-like morphology line that was isolated from the liver of a female rat with hepatoma. This cell line was deposited by JE Becker and can be used in cancer research.
Catalog No. 50-238-2497