Learn More
Cayman Chemical McaRPPGFSAFK DnpOHtrIflu 10mg
SDP
Catalog No. 502915283
Encompass_Preferred
A fluorogenic substrate for ECE-1; Mca is released upon cleavage by ECE-1 and its fluorescence can be used to quantify ECE-1 activity; ex/em = 328/420 nm, respectively
Catalog No. 50-291-5283 Supplier Cayman Chemical Supplier No. 3688710 MG
A fluorogenic substrate for ECE-1; Mca is released upon cleavage by ECE-1 and its fluorescence can be used to quantify ECE-1 activity; ex/em = 328/420 nm, respectively