Learn More
Aviva Systems Biology Corp MCART6 Antibody - middle region (ARP44642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCART6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MCART6. Alternate names: DKFZp686O1267, MCART6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-63053