Learn More
Cayman Chemical McaSEVNLDAEFRK DnpRRNH2 10mg
SDP
Catalog No. 502914901
Encompass_Preferred
A fluorogenic substrate for BACE; Mca is released upon cleavage by BACE and its fluorescence can be used to quantify BACE activity; ex/em = 328/420 nm, respectively
Catalog No. 50-291-4901 Supplier Cayman Chemical Supplier No. 3687910 MG
A fluorogenic substrate for BACE; Mca is released upon cleavage by BACE and its fluorescence can be used to quantify BACE activity; ex/em = 328/420 nm, respectively