Learn More
Medchemexpress LLC MCB-22-174 | 3058199-58-6 | 99.8% | 429.37 | 5 MG
SDP
Catalog No. 5000394232
Encompass_Preferred
MCB-22-174 is a deuterated Piezo1 agonist with an EC50 value of 6.28 μM. It activates the CaMKII/ERK signaling pathway and initiates Ca2+ influx in rMSCs. It decreases the expression of chondrogenesis markers (Comp, Acan) and adipogenesis markers (Lpl, Fabp4) in MSCs. This compound can improve bone quality in hind-limb unloading (HU) model rats and can be used for the study of disuse osteoporosis (OP).
  • Deuterated Piezo1 agonist
  • Activates the CaMKII/ERK signaling pathway
  • Initiates Ca2+ influx in rMSCs
  • Decreases expression of chondrogenesis markers (Comp, Acan) in MSCs
  • Decreases expression of adipogenesis markers (Lpl, Fabp4) in MSCs
  • Effectively improves bone quality in hind-limb unloading (HU) model rats
  • Can be used for the study of disuse osteoporosis (OP)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-94232 Supplier Medchemexpress LLC Supplier No. HY165607S5MG
MCB-22-174 is a deuterated Piezo1 agonist with an EC50 value of 6.28 μM. It activates the CaMKII/ERK signaling pathway and initiates Ca2+ influx in rMSCs. It decreases the expression of chondrogenesis markers (Comp, Acan) and adipogenesis markers (Lpl, Fabp4) in MSCs. This compound can improve bone quality in hind-limb unloading (HU) model rats and can be used for the study of disuse osteoporosis (OP).
  • Deuterated Piezo1 agonist
  • Activates the CaMKII/ERK signaling pathway
  • Initiates Ca2+ influx in rMSCs
  • Decreases expression of chondrogenesis markers (Comp, Acan) in MSCs
  • Decreases expression of adipogenesis markers (Lpl, Fabp4) in MSCs
  • Effectively improves bone quality in hind-limb unloading (HU) model rats
  • Can be used for the study of disuse osteoporosis (OP)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS