Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MCB-3681 | 790704-42-6 | 99.2% | 626.60 | 10 MG
SDP

Catalog No. 5000438892
Encompass_Preferred

MCB-3681 is the active substance of the antibacterial Oxaquin, effective against gram-positive bacterium. It is used in laboratory chemicals and for the manufacture of substances. This product is for research use only and intended for experienced personnel.

  • Appearance: Solid, light yellow to yellow
  • Stable under recommended storage conditions
  • Not a hazardous substance or mixture
  • Avoid inhalation, contact with eyes and skin
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 62.5 mg/mL (99.74 mM)
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.08 mg/mL)
  • Soluble in 10% DMSO, 90% Corn Oil (≥ 2.08 mg/mL)

Catalog No. 50-004-38892 Supplier Medchemexpress LLC Supplier No. HY11190210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MCB-3681 is the active substance of the antibacterial Oxaquin, effective against gram-positive bacterium. It is used in laboratory chemicals and for the manufacture of substances. This product is for research use only and intended for experienced personnel.

  • Appearance: Solid, light yellow to yellow
  • Stable under recommended storage conditions
  • Not a hazardous substance or mixture
  • Avoid inhalation, contact with eyes and skin
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO: 62.5 mg/mL (99.74 mM)
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.08 mg/mL)
  • Soluble in 10% DMSO, 90% Corn Oil (≥ 2.08 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.