Learn More
Medchemexpress LLC MCB-613 | 1162656-22-5 | 99.6% | 304.39 | 1 ML
SDP
Catalog No. 5000402790
Encompass_Preferred
MCB-613 is a potent small molecule stimulator of Steroid Receptor Coactivators (SRC), designed to super-stimulate their transcriptional activity. It enhances SRCs' interactions with other coactivators and notably induces ER stress linked to the generation of reactive oxygen species (ROS). Targeting oncogenes, MCB-613 acts as an anti-cancer agent by over-stimulating the SRC oncogenic program. This product is for research use only.
  • Activates endogenous MMP13 mRNA expression.
  • Induces proteasome dysfunction and ER stress.
  • Triggers markers for unfolded protein response (UPR).
  • Impacts SRC-3 KO and WT HeLa cell viability.
  • Stalls tumor growth in MCF-7 breast cancer mouse xenograft models.
Catalog No. 50-004-02790 Supplier Medchemexpress LLC Supplier No. HY1962510MM/1ML
MCB-613 is a potent small molecule stimulator of Steroid Receptor Coactivators (SRC), designed to super-stimulate their transcriptional activity. It enhances SRCs' interactions with other coactivators and notably induces ER stress linked to the generation of reactive oxygen species (ROS). Targeting oncogenes, MCB-613 acts as an anti-cancer agent by over-stimulating the SRC oncogenic program. This product is for research use only.
  • Activates endogenous MMP13 mRNA expression.
  • Induces proteasome dysfunction and ER stress.
  • Triggers markers for unfolded protein response (UPR).
  • Impacts SRC-3 KO and WT HeLa cell viability.
  • Stalls tumor growth in MCF-7 breast cancer mouse xenograft models.