Learn More
Apexbio Technology LLC MCB-613, 25mg. Cas: 1162656-22-5 MFCD: NA. stimulator of steroid receptor coactivator (SRC)
SDP
Supplier:  Apexbio Technology LLC A871225
Encompass_Preferred
MCB-613, 4-ethyl-2,6-bis-pyridin-3-ylmethylene-cyclohexanone, is a novel and potent stimulator of p160 steroid receptor coactivators (SRCs). It is a Pan-SRCs stimulator. MCB-613 is conrmed to be a strong activator of all three SRCs family proteins [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5181