Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MCC-AAQ-Exa | 3052836-65-1 | 95.3% | 924.97 | C47H53FN8O11 | 5 MG
SDP

Catalog No. 5000202756
Encompass_Preferred

MCC-AAQ-Exa is a drug-linker conjugate used in the development of antibody-drug conjugates (ADCs). It is produced by condensing an MCC maleimide-containing linker with an exatecan payload and is supplied as a research reagent for conjugation workflows.

  • Drug-linker conjugate for antibody-drug conjugate (ADC) development.
  • Contains an MCC maleimide linker enabling thiol conjugation to antibodies.
  • Derived from exatecan payload for potent cytotoxic activity once internalized.
  • Molecular weight 924.97.
  • Chemical formula C47H53FN8O11.
  • Purity 95.3% (as supplied).
  • CAS number 3052836-65-1.
  • Recommended storage: -20°C, protect from light, stored under inert gas; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-02756 Supplier Medchemexpress LLC Supplier No. HY1742795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MCC-AAQ-Exa is a drug-linker conjugate used in the development of antibody-drug conjugates (ADCs). It is produced by condensing an MCC maleimide-containing linker with an exatecan payload and is supplied as a research reagent for conjugation workflows.

  • Drug-linker conjugate for antibody-drug conjugate (ADC) development.
  • Contains an MCC maleimide linker enabling thiol conjugation to antibodies.
  • Derived from exatecan payload for potent cytotoxic activity once internalized.
  • Molecular weight 924.97.
  • Chemical formula C47H53FN8O11.
  • Purity 95.3% (as supplied).
  • CAS number 3052836-65-1.
  • Recommended storage: -20°C, protect from light, stored under inert gas; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.