Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nicotinate, Reagent, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604428 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-428 12 kg Amber Glass Bottle
18-604-425 100 g Amber Glass Bottle
18-604-426 500 g Amber Glass Bottle
18-604-427 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-428 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M127612KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1276, 93-60-7, C7H7NO2

Spectrum™ Chemical Methyl Nicotinate, Reagent

Chemical Identifiers

CAS 93-60-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key YNBADRVTZLEFNH-UHFFFAOYSA-N
IUPAC Name methyl pyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1

Specifications

CAS Max % 100%
Boiling Point 209(°C) (408.2 (°F)
Packaging Amber Glass Bottle
Quantity 12 kg
Percent Purity 99%
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Methyl Nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.