Learn More
Selleck Chemical LLC MCC950 Sodium (CP-456773 Sodium) 50mg 256373-96-3 CRID3 sodium salt
SDP
Supplier:  Selleck Chemical LLC S780950MG
Encompass_Preferred
MCC950 Sodium (CP-456773 Sodium, CRID3 sodium salt) is a potent, selective inhibitor of NLRP3 with IC50 of 7.5 nM in BMDMs; but not the AIM2, NLRC4 or NLRP1 inflammasomes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5283