Learn More
TARGETMOL CHEMICALS INC MCC950 sodium
SDP
Supplier:  TARGETMOL CHEMICALS INC T6887100MG
Encompass_Preferred
MCC950 sodium salt (IC50 of 7.5 nM in BMDMs) is an effective, selective NLRP3 inhibitor.
Catalog No. NC1727148