Learn More
Medchemexpress LLC MCC950 sodium | 256373-96-3 | 99.8% | 200 MG
SDP
Catalog No. 5000339680
Encompass_Preferred
MCC950 sodium is a potent and selective NLRP3 inhibitor with IC50s of 7.5 nM in BMDMs and 8.1 nM in HMDMs. It blocks canonical and non-canonical NLRP3 activation at nanomolar concentrations and specifically inhibits NLRP3 but not AIM2, NLRC4, or NLRP1 activation.
  • Potent and selective NLRP3 inhibitor
  • Inhibits IL-1β but not TNF-α secretion
  • Blocks canonical and non-canonical NLRP3 activation
  • Specifically inhibits NLRP3 (not AIM2, NLRC4, or NLRP1)
  • Reduces Interleukin-1β (IL-1β) production
  • Attenuates severity of experimental autoimmune encephalomyelitis (EAE)
  • Off-white to yellow solid appearance
  • Purity of 99.8%
Catalog No. 50-003-39680 Supplier Medchemexpress LLC Supplier No. HY12815A200MG
MCC950 sodium is a potent and selective NLRP3 inhibitor with IC50s of 7.5 nM in BMDMs and 8.1 nM in HMDMs. It blocks canonical and non-canonical NLRP3 activation at nanomolar concentrations and specifically inhibits NLRP3 but not AIM2, NLRC4, or NLRP1 activation.
  • Potent and selective NLRP3 inhibitor
  • Inhibits IL-1β but not TNF-α secretion
  • Blocks canonical and non-canonical NLRP3 activation
  • Specifically inhibits NLRP3 (not AIM2, NLRC4, or NLRP1)
  • Reduces Interleukin-1β (IL-1β) production
  • Attenuates severity of experimental autoimmune encephalomyelitis (EAE)
  • Off-white to yellow solid appearance
  • Purity of 99.8%