Learn More
Medchemexpress LLC HY-12815A 50mg Medchemexpress, MCC950 (sodium) CAS:256373-96-3 Purity:>98%
SDP
Catalog No. 502028918
Encompass_Preferred
Medchemexpress, HY-12815A 50mg MCC950 (sodium) CAS:256373-96-3 MCC950 sodium (CP-456773 sodium; CRID3 sodium salt) is a potent, selective NLRP3 inhibitor with IC50s of 7.5 and 8.1 nM in BMDMs and HMDMs, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8918 Supplier Medchemexpress LLC Supplier No. HY12815A50MG
Medchemexpress, HY-12815A 50mg MCC950 (sodium) CAS:256373-96-3 MCC950 sodium (CP-456773 sodium; CRID3 sodium salt) is a potent, selective NLRP3 inhibitor with IC50s of 7.5 and 8.1 nM in BMDMs and HMDMs, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.