Learn More
ATCC MC/CAR; Plasmacytoma; Human
Catalog No. 502383369
Encompass
MC/CAR is a lymphoblast-like cell that was isolated from the peripheral blood of an 81-year-old, White male with plasmacytoma. The cell can be used in immune system disorders research and as a human fusion line.
Catalog No. 50-238-3369 Supplier ATCC Supplier No. CRL8083
Add to Cart
Add to Cart
MC/CAR is a lymphoblast-like cell that was isolated from the peripheral blood of an 81-year-old, White male with plasmacytoma. The cell can be used in immune system disorders research and as a human fusion line.