Learn More
ABclonal Technology MCCC2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15181500UL
Encompass_Preferred
This gene encodes the small subunit of 3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoA to form 3-methylglutaconyl-CoA. Mutations in this gene are associated with 3-Methylcrotonylglycinuria, an autosomal recessive disorder of leucine catabolism. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Catalog No. 50-001-09213