Learn More
Bioworld McCown and Murashige and Skoog (MS) Vitamin Concentration (Syringe Vials), 100 mL
Catalog No. 50254653
Encompass_Preferred
McCown and Murashige & Skoog (MS) Vitamin ConcentrationContains Vitamins as described by Lloyd & McCown (1981) and Murashige & Skoog (1962)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: Glycine: 2.00 mg/Lmyo-Inositol: 100.00 mg/L Nicotinic Acid: 0.50 mg/L Pyridoxine HCl: 0.50 mg/LThiamine HCl: 1.00 mg/L Total: 2462.60 mg/L
Catalog No. 50-254-653 Supplier Bioworld Supplier No. 306300521
McCown and Murashige & Skoog (MS) Vitamin ConcentrationContains Vitamins as described by Lloyd & McCown (1981) and Murashige & Skoog (1962)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: Glycine: 2.00 mg/Lmyo-Inositol: 100.00 mg/L Nicotinic Acid: 0.50 mg/L Pyridoxine HCl: 0.50 mg/LThiamine HCl: 1.00 mg/L Total: 2462.60 mg/L