Learn More
Bioworld McCown and Murashige and Skoog (MS) Vitamin Mix, 100 mL
Catalog No. 50254654
Encompass_Preferred
McCown and Murashige & Skoog (MS) Vitamin MixContains Vitamins as described by Lloyd & McCown (1981) and Murashige & Skoog (1962) Package contains 10.40 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: Glycine: 2.00 mg/L myo-Inositol: 100.00 mg/L Nicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/LThiamine HCl: 1.00 mg/L
Catalog No. 50-254-654 Supplier Bioworld Supplier No. 306300531
McCown and Murashige & Skoog (MS) Vitamin MixContains Vitamins as described by Lloyd & McCown (1981) and Murashige & Skoog (1962) Package contains 10.40 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: Glycine: 2.00 mg/L myo-Inositol: 100.00 mg/L Nicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/LThiamine HCl: 1.00 mg/L