Learn More
Research Products International Corp McCown's Woody Plant Medium, Powder, Makes 25 Liters
SDP
Catalog No. 50213417
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Lloyd & McCown (1980).
Catalog No. 50-213-417 Supplier Research Products International Corp Supplier No. M1010025.0
Basal salt mixture containing micro and macro elements as described by Lloyd & McCown (1980).