Learn More
Bioworld McCown Woody Plant Basal Salt Mixture, 50 L
Catalog No. 50254663
Encompass_Preferred
McCown Woody Plant Basal Salt MixtureBasal salt mixture containing Micronutrients and Macronutrients as described by Lloyd & McCown (1981)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.30 g/LpH: 3.5–4.5Soluble in waterUsually ships within 24 hours This product is sold for laboratory research use only
Catalog No. 50-254-663 Supplier Bioworld Supplier No. 306300564
McCown Woody Plant Basal Salt MixtureBasal salt mixture containing Micronutrients and Macronutrients as described by Lloyd & McCown (1981)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.30 g/LpH: 3.5–4.5Soluble in waterUsually ships within 24 hours This product is sold for laboratory research use only