Learn More
Bioworld McCown Woody Plant Basal Salt Mixture w/ Vitamins, 25 L
Catalog No. 50254665
Encompass_Preferred
McCown's Woody Plant Basal Salt MixtureBasal salt mixture containing Micronutrients, Macronutrients, and Vitamins as described by Lloyd & McCown (1981)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.41 g/LpH: 3.5−4.5Soluble in waterUsually ships within 24 hours This product is sold for laboratory research use only
Catalog No. 50-254-665 Supplier Bioworld Supplier No. 306300573
McCown's Woody Plant Basal Salt MixtureBasal salt mixture containing Micronutrients, Macronutrients, and Vitamins as described by Lloyd & McCown (1981)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 2.41 g/LpH: 3.5−4.5Soluble in waterUsually ships within 24 hours This product is sold for laboratory research use only