Learn More
Research Products International Corp McCown's Woody Plant Medium with Vitamins, Powder, Makes 1 Liter
SDP
Catalog No. 50213453
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Lloyd & McCown (1980).
Catalog No. 50-213-453 Supplier Research Products International Corp Supplier No. M230001.0
Basal salt mixture containing micro and macro elements with vitamins as described by Lloyd & McCown (1980).