Learn More
ATHENA ES McCoy's 5A
Catalog No. 5000158135
Encompass_Preferred
McCoy's Medium 5A is a general medium for primary and established cell lines. It contains increased glucose, additional vitamins, high phosphate, and peptone.
Catalog No. 50-001-58135 Supplier ATHENA ES Supplier No. 0466
McCoy's Medium 5A is a general medium for primary and established cell lines. It contains increased glucose, additional vitamins, high phosphate, and peptone.