Learn More
ATCC McCoy's 5A Medium, 500mL
Supplier:  ATCC 302007/DEL
Encompass
McCoy's 5A Medium is modified to contain 1.5 mM L-glutamine and 2200 mg/L sodium bicarbonate. NOTE: This reduced level of sodium bicarbonate (NaHCO3, 2.2 g/L) is intended for use in 5% CO2 in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO2. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188272NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-272FP