Learn More
ATCC McCoy's 5A Medium
Supplier:  ATCC 302007
Encompass
McCoy's 5A Medium is modified to contain 1.5 mM L-glutamine and 2200 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3814