Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc McCoys 5A Medium with L-Glutamine 500 mL
SDP

Catalog No. 502209730
Encompass_Preferred

McCoys 5A Medium is a general purpose medium that supports the propagation of many types of primary cells established cell lines and explants from biopsy tissues. McCoys 5A Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

With L-Glutamine
With Phenol Red

Sterile Filtered
pH: 7.0±0.2
Osmolality: 290±5% mOsm/Kg H2O
Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9730 Supplier Quality Biological Inc Supplier No. 112028101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

McCoys 5A Medium is a general purpose medium that supports the propagation of many types of primary cells established cell lines and explants from biopsy tissues. McCoys 5A Medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

With L-Glutamine
With Phenol Red

Sterile Filtered
pH: 7.0±0.2
Osmolality: 290±5% mOsm/Kg H2O
Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.