Learn More
ILEX LIFE SCIENCES LLC MCCOYS 5A P04-05500 6 X 500 ML
SDP
Catalog No. 502733873
Encompass_Preferred
PAN-Biotech McCoy’s 5A Medium (modified), w: L-Glutamine, w: 2.2 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: McCoy's 5A Medium (modified) is a complete medium with all amino acids and vitamins. It is used for growing primary cultures. This group contains marrow cells, gingival cells, adrenal cells, spleen cells, lung cells, rat embryos and other cell types. McCoy's 5A medium was originally developed by Dr. Thomas McCoy as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A is unique in that it contains the reducing agent glutathione, bacto-peptone, and a high level of glucose.
Catalog No. 50-273-3873 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0405500X6
Add to Cart
Add to Cart
PAN-Biotech McCoy’s 5A Medium (modified), w: L-Glutamine, w: 2.2 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: McCoy's 5A Medium (modified) is a complete medium with all amino acids and vitamins. It is used for growing primary cultures. This group contains marrow cells, gingival cells, adrenal cells, spleen cells, lung cells, rat embryos and other cell types. McCoy's 5A medium was originally developed by Dr. Thomas McCoy as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A is unique in that it contains the reducing agent glutathione, bacto-peptone, and a high level of glucose.