Learn More
FUJIFILM Biosciences McCoys Medium 5A - Liquid
Catalog No. 5000450300
Encompass
McCoy s 5A medium is a modification of Basal Medium Eagle BME with changes in amino acid compositions to support optimal hepatoma cultures Containing additional vitamins high phosphate and Bacto peptone McCoy s 5A medium has been demonstrated to support primary cell cultures from many normal adult tissues including bone marrow testes kidney skin and gingiva
Catalog No. 50-004-50300 Supplier FUJIFILM Biosciences Supplier No. 9090500ML
McCoy s 5A medium is a modification of Basal Medium Eagle BME with changes in amino acid compositions to support optimal hepatoma cultures Containing additional vitamins high phosphate and Bacto peptone McCoy s 5A medium has been demonstrated to support primary cell cultures from many normal adult tissues including bone marrow testes kidney skin and gingiva