Learn More
Sigma Aldrich Fine Chemicals Biosciences MCDB 105 Medium, 500 mL
Catalog No. NC1905831
Encompass
All-in-one ready-to-use media. Each lot was tested for the ability to support plating, spreading and proliferation.
Catalog No. NC1905831 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 117500
All-in-one ready-to-use media. Each lot was tested for the ability to support plating, spreading and proliferation.