Learn More
Sigma Aldrich Fine Chemicals Biosciences MCDB 153 Medium, 1 L
Catalog No. NC1225423
Encompass
With trace elements, L-glutamine and 28mM HEPES, without sodium bicarbonate, powder, suitable for cell culture. Originally developed for clonal and long term growth of human epidermal keratinocytes under serum-free conditions.
Catalog No. NC1225423 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M74031L
With trace elements, L-glutamine and 28mM HEPES, without sodium bicarbonate, powder, suitable for cell culture. Originally developed for clonal and long term growth of human epidermal keratinocytes under serum-free conditions.