Learn More
StatLab McDonald's Gram MasterTech Special Stain Kit
SDP
Catalog No. 22000052
Change view
Click to view available options
100 mL
Catalog No.
22-000-052 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-052 Supplier StatLab Supplier No. KTMGS
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for gram positive and gram negative bacteria in less than five minutes
  • Both Gentian Violet and Carbol Fuchsin serve as the primary stains, producing blue gram positive bacteria and red gram negative bacteria against a yellow background, counterstained by Optimized Tartrazine which provides more intense staining in less time than traditional Tartrazine formulations
  • 100 mL Stain Kit yields between 36 and 45 slides
  • A MasterTech Special Stain Kit
Specifications
Product Type Gram Decolorizing Solution