Learn More
CELLTREAT Scientific Products MCE Syringe Filter, 0.22m, 13mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229750
Encompass_Preferred
CELLTREAT Scientific Products 0.22¿m MCE Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. MCE (Mixed Cellulose Ester). Filtration of aqueous solutions. Effectively binds trace proteins. Oil particulates and bacteria filtration. Membrane diameter 13mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in peel-to-open paper/plastic blister packs. 75/case.
Catalog No. 50-202-065