Learn More
WELCH MATERIALS INC Membrane filter, MCE, Dia. 47mm, Pore: 0.45µm, Grid, Sterile, individually packed. 100pcs/pk, 10pk/cs.
Catalog No. NC2052802
Encompass_Preferred
Membrane filter, MCE, Dia. 47mm, Pore: 0.45µm, Grid, Sterile, individually packed. 100pcs/pk, 10pk/cs.
Catalog No. NC2052802 Supplier WELCH MATERIALS INC Supplier No. M47MCE045GWS10
Add to Cart
Add to Cart
Membrane filter, MCE, Dia. 47mm, Pore: 0.45µm, Grid, Sterile, individually packed. 100pcs/pk, 10pk/cs.